4-Thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T2494-1G T2494-5G
TCT2494-1GEA 113.76 CAD
TCT2494-1G TCT2494-5G
4-Thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
4-Thiazolecarboxylic acid
Formula: C₄H₃NO₂S
MDL Number: MFCD00623589
CAS Number: 3973-08-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR