(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

A2830-1G A2830-5G
TCA2830-1GEA 186.44 CAD
TCA2830-1G TCA2830-5G
(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
(R)-(+)-3-Aminoquinuclidine dihydrochloride
Formula: C₇H₁₆Cl₂N₂
Melting Pt: 315°C
MDL Number: MFCD00191752
CAS Number: 123536-14-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +22.0 to +26.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR