5-Methylfuran-2-boronic acid pinacol ester ≥97.0% (by GC, titration analysis)

Supplier: TCI America

T3582-1G T3582-5G
TCT3582-1GEA 44.24 CAD
TCT3582-1G TCT3582-5G
5-Methylfuran-2-boronic acid pinacol ester ≥97.0% (by GC, titration analysis)
5-Methylfuran-2-boronic acid pinacol ester
Formula: C₁₁H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03094689
CAS Number: 338998-93-9

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR