tert-Butyl (dimethylphosphono)acetate ≥95.0% (by GC)

Supplier: TCI America
Warning

B5094-25G B5094-5G
TCB5094-25GEA 171.9 CAD
TCB5094-25G TCB5094-5G
tert-Butyl (dimethylphosphono)acetate ≥95.0% (by GC)
tert-Butyl (dimethylphosphono)acetate
Formula: C₈H₁₇O₅P
MDL Number: MFCD00042939
CAS Number: 62327-21-3

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR