Morpholin-3-one ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Morpholinone

M2788-25G M2788-5G
TCM2788-25GEA 314.42 CAD
TCM2788-25G TCM2788-5G
Morpholin-3-one ≥98.0% (by GC)
Morpholin-3-one
Formula: C₄H₇NO₂
Boiling Pt: 142°C
MDL Number: MFCD00631009
CAS Number: 109-11-5
EINECS: 203-647-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 103.0 to 107.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR