2,3,5-Tribromopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

T3339-25G T3339-5G
76220-566EA 238.58 CAD
76220-566 76220-568
2,3,5-Tribromopyridine ≥98.0% (by GC)
2,3,5-Tribromopyridine
Formula: C₅H₂Br₃N
Boiling Pt: 160°C
MDL Number: MFCD00233999
CAS Number: 75806-85-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR