Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)

Supplier: TCI America

F0998-25G F0998-5G
TCF0998-25GEA 371.3 CAD
TCF0998-25G TCF0998-5G
Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)
Ferrocenecarbaldehyde
Formula: C₁₁H₁₀FeO
MDL Number: MFCD00001429
CAS Number: 12093-10-6
EINECS: 235-158-1

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Purity (Chelometric Titration) min. 97.0 %
Melting point 114.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR