3,5-Dichloro-4-iodopyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D5195-1G
D5195-5G
TCD5195-1GEA
63.2
CAD
TCD5195-1G
TCD5195-5G
3,5-Dichloro-4-iodopyridine ≥98.0% (by GC, titration analysis)
3,5-Dichloro-4-iodopyridine
Formula:
C₅H₂Cl₂IN |
MDL Number:
MFCD03094687 CAS Number: 343781-41-9 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...