2,6-Dichloro-4-iodopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D4774-1G D4774-5G
TCD4774-1GEA 104.28 CAD
TCD4774-1G TCD4774-5G
2,6-Dichloro-4-iodopyridine ≥98.0% (by GC)
2,6-Dichloro-4-iodopyridine
Formula: C₅H₂CI₂IN
MDL Number: MFCD07368400
CAS Number: 98027-84-0

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR