1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥98.0% (by GC)

Supplier: TCI America
Danger

H1507-25G H1507-5G
TCH1507-25GEA 532.3 CAD
TCH1507-25G TCH1507-5G
1,1,2,3,3,3-Hexafluoropropyl methyl ether ≥98.0% (by GC)
1,1,2,3,3,3-Hexafluoropropyl methyl ether
Formula: C₄H₄F₆O
Boiling Pt: 55°C
MDL Number: MFCD00041537
CAS Number: 382-34-3
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR