1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid Pinacol ester, 1-(Tetrahydropyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester, 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
T3215-1G
T3215-5G
TCT3215-1GEA
33.18
CAD
TCT3215-1G
TCT3215-5G
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester ≥98.0% (by GC, titration analysis)
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
Formula:
C₁₄H₂₃BN₂O₃ MW: 278.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09037501 CAS Number: 903550-26-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...