Resorcinol monobenzoate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Benzoic acid 3-Hydroxyphenyl ester, 3-Hydroxyphenyl Benzoate, Resorcinol monobenzoate

H1560-100G H1560-25G
TCH1560-100GEA 42.66 CAD
TCH1560-100G TCH1560-25G
Resorcinol monobenzoate ≥95.0% (by GC)
Resorcinol monobenzoate
Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
MDL Number: MFCD00020118
CAS Number: 136-36-7
EINECS: 205-241-7

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 131.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR