Piperidine-2,4-dione ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,4-Piperidinedione, 2,4-Dioxopiperidine

P2296-1G P2296-200MG
TCP2296-1GEA 96.38 CAD
TCP2296-1G TCP2296-200MG
Piperidine-2,4-dione ≥98.0% (by GC, titration analysis)
Piperidine-2,4-dione
Formula: C₅H₇NO₂
MW: 113.1146 g/mol
MDL Number: MFCD08704814
CAS Number: 50607-30-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR