2-Amino-6-methylpyrazine ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Methylpyrazin-2-amine

A2992-1G A2992-200MG
TCA2992-1GEA 148.52 CAD
TCA2992-1G TCA2992-200MG
2-Amino-6-methylpyrazine ≥97.0% (by GC)
2-Amino-6-methylpyrazine
Formula: C₅H₇N₃
MDL Number: MFCD08235193
CAS Number: 5521-56-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR