2-Ethyl-6-methylphenol ≥98.0% (by GC)

Supplier: TCI America
Warning

E1185-1G E1185-5G
TCE1185-1GEA 66.36 CAD
TCE1185-1G TCE1185-5G
2-Ethyl-6-methylphenol ≥98.0% (by GC)
2-Ethyl-6-methylphenol
Formula: C₉H₁₂O
Boiling Pt: 214°C
MDL Number: MFCD00962292
CAS Number: 1687-64-5

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR