Tripalmitin (glycerol tripalmitate) ≥80.0% (by GC)

Supplier: TCI America

Synonyms: Propane-1,2,3-triyl tripalmitate, Tripalmitin, Glycerol tripalmitate

T3096-100G T3096-25G
TCT3096-100GEA 410.8 CAD
TCT3096-100G TCT3096-25G
Tripalmitin (glycerol tripalmitate) ≥80.0% (by GC)
Tripalmitin (Glycerol tripalmitate)
Formula: C₅₁H₉₈O₆
MW: 807.34 g/mol
Boiling Pt: 320°C
MDL Number: MFCD00008995
CAS Number: 555-44-2
EINECS: 209-098-1
Merck Index: 14,09736

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 80.0 %
Melting point 63.0 to 68.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR