7-(Ethylamino)-4,6-dimethylcoumarin ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
E1132-1G
E1132-200MG
TCE1132-1GEA
289.3
CAD
TCE1132-1G
TCE1132-200MG
7-(Ethylamino)-4,6-dimethylcoumarin ≥98.0% (by GC)
7-(Ethylamino)-4,6-dimethylcoumarin
Formula:
C₁₃H₁₅NO₂ |
MDL Number:
MFCD00006860 CAS Number: 26078-25-1 EINECS: 247-446-4 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 169.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...