L(-)-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C3058-25G C3058-5G
TCC3058-25GEA 210.46 CAD
TCC3058-25G TCC3058-5G
L(-)-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)
L(-)-Carnitine hydrochloride
Formula: C₇H₁₆ClNO₃
MDL Number: MFCD00066100
CAS Number: 6645-46-1
EINECS: 229-663-6
Merck Index: 14,01849

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C
Specific rotation [a]20/D -21.0 to -25.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR