2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5076-1G D5076-5G
76219-828EA 75.84 CAD
76219-828 76219-830
2,4-Dichloro-3-formylpyridine ≥98.0% (by GC)
2,4-Dichloro-3-formylpyridine
Formula: C₆H₃Cl₂NO
MDL Number: MFCD07437909
CAS Number: 134031-24-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR