1-(4-Chlorophenyl)cyclobutanecarbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

C3161-25G C3161-5G
76218-910EA 1290.86 CAD
76218-910 76218-912
1-(4-Chlorophenyl)cyclobutanecarbonitrile ≥98.0% (by GC)
1-(4-Chlorophenyl)cyclobutanecarbonitrile
Formula: C₁₁H₁₀ClN
Boiling Pt: 112°C
MDL Number: MFCD00065239
CAS Number: 28049-61-8
EINECS: 248-799-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR