Benzo[b]thiophene-2-carbonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B4637-1G B4637-5G
TCB4637-1GEA 134.3 CAD
TCB4637-1G TCB4637-5G
Benzo[b]thiophene-2-carbonyl chloride ≥98.0% (by titrimetric analysis)
Benzo[b]thiophene-2-carbonyl chlorid
Formula: C₉H₅ClOS
Boiling Pt: 176°C
MDL Number: MFCD00051635
CAS Number: 39827-11-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 83.0 to 88.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR