3-Phosphonopropionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 3-Phosphonopropanoic acid

P2348-1G P2348-5G
TCP2348-1GEA 69.36 CAD
TCP2348-1G TCP2348-5G
3-Phosphonopropionic acid ≥98.0% (by titrimetric analysis)
3-Phosphonopropionic acid
Formula: C₃H₇O₅P
MDL Number: MFCD00002770
CAS Number: 5962-42-5
EINECS: 227-738-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR