Ethephon(2-chloroethylphosphonic acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
C1456-1G
C1456-5G
TCC1456-1GEA
194.97
CAD
TCC1456-1G
TCC1456-5G
Ethephon(2-chloroethylphosphonic acid) ≥98.0% (by titrimetric analysis)
Ethephon (2-Chloroethylphosphonic acid)
Formula:
C₂H₆ClO₃P Boiling Pt: 265°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002142 CAS Number: 16672-87-0 EINECS: 240-718-3 UN: 2923 ADR: 8,II Merck Index: 14,03732 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 73.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...