Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)

Supplier: TCI America
Warning

D3071-25G D3071-5G
TCD3071-25GEA 237 CAD
TCD3071-25G TCD3071-5G
Diethyl (bromodifluoromethyl)phosphonate ≥97.0% (by GC)
Diethyl (bromodifluoromethyl)phosphonate
Formula: C₅H₁₀BrF₂O₃P
Boiling Pt: 102°C
MDL Number: MFCD00069118
CAS Number: 65094-22-6

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR