4,6-Dichloro-5-methoxypyrimidine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D4723-25G
D4723-5G
TCD4723-25GEA
171.9
CAD
TCD4723-25G
TCD4723-5G
4,6-Dichloro-5-methoxypyrimidine ≥98.0% (by GC)
4,6-Dichloro-5-methoxypyrimidine
Formula:
C₅H₄Cl₂N₂O MW: 179.01 g/mol |
MDL Number:
MFCD00087680 CAS Number: 5018-38-2 EINECS: 225-699-1 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...