4,6-Dichloro-2-methylquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

D4782-1G D4782-5G
TCD4782-1GEA 194.34 CAD
TCD4782-1G TCD4782-5G
4,6-Dichloro-2-methylquinoline ≥98.0% (by GC)
4,6-Dichloro-2-methylquinoline
Formula: C₁₀H₇Cl₂N
MDL Number: MFCD00272372
CAS Number: 53342-53-3

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR