7-Chloroquinoline ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3085-25G
C3085-5G
TCC3085-25GEA
1058.6
CAD
TCC3085-25G
TCC3085-5G
7-Chloroquinoline ≥98.0% (by GC)
7-Chloroquinoline
Formula:
C₉H₆ClN Boiling Pt: 268°C Storage Temperature: Refrigerator |
MDL Number:
MFCD00956370 CAS Number: 612-61-3 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...