1-Ethyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

E1307-25G E1307-5G
76220-054EA 546.68 CAD
76220-054 76220-056
1-Ethyl-3-methylimidazoliumtrifluoroacetate ≥97.0% (by titrimetric analysis)
1-Ethyl-3-methylimidazolium trifluoroacetate
Formula: C₈H₁₁F₃N₂O₂
MDL Number: MFCD08458492
CAS Number: 174899-65-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR