(4-Bromobenzyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B5241-25G
B5241-5G
76219-538EA
285.98
CAD
76219-538
76219-540
(4-Bromobenzyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(4-Bromobenzyl)triphenylphosphonium bromide
Formula:
C₂₅H₂₁Br₂P Melting Pt: 275°C |
MDL Number:
MFCD00051922 CAS Number: 51044-13-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...