1-Allyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A2084-25G A2084-5G
76218-734EA 242.69 CAD
76218-734 76218-736
1-Allyl-3-methylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Allyl-3-methylimidazolium chloride
Formula: C₇H₁₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD07784450
CAS Number: 65039-10-3

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR