1-(2-Hydroxyethyl)-3-methylimidazolium chloride ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
H1254-25G
H1254-5G
76220-116EA
295.46
CAD
76220-116
76220-118
1-(2-Hydroxyethyl)-3-methylimidazolium chloride ≥98.0% (by HPLC)
1-(2-Hydroxyethyl)-3-methylimidazolium chloride
Formula:
C₆H₁₁ClN₂O |
MDL Number:
MFCD12761437 CAS Number: 61755-34-8 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 82.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...