(3,3,3-Trifluoropropyl)dichloromethylsilane ≥98.0% (by GC)

Supplier: TCI America
Danger

D4729-25G D4729-5G
TCD4729-25GEA 133.45 CAD
TCD4729-25G TCD4729-5G
(3,3,3-Trifluoropropyl)dichloromethylsilane ≥98.0% (by GC)
(3,3,3-Trifluoropropyl)dichloromethylsilane
Formula: C₄H₇Cl₂F₃Si
Boiling Pt: 123°C
Flash Pt: 15°C
MDL Number: MFCD00039294
CAS Number: 675-62-7
EINECS: 211-623-4
UN: 2985
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR