Hygromycin B ≥80.0% (by HPLC)

Supplier: TCI America
Danger

H1509-1G H1509-200MG
TCH1509-1GEA 290.88 CAD
TCH1509-1G TCH1509-200MG
Hygromycin B ≥80.0% (by HPLC)
Hygromycin B
Formula: C₂₀H₃₇N₃O₁₃
Melting Pt: 180°C
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I
Merck Index: 14,04852

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) (CAD) min. 80.0 area%
Specific rotation [a]20/D +18.0 to +23.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR