4-Benzyloxyphenylacetonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(4-(Benzyloxy)phenyl)acetonitrile

B4734-1G B4734-5G
TCB4734-1GEA 66.36 CAD
TCB4734-1G TCB4734-5G
4-Benzyloxyphenylacetonitrile ≥98.0% (by GC)
4-Benzyloxyphenylacetonitrile
Formula: C₁₅H₁₃NO
Boiling Pt: 173°C
MDL Number: MFCD00016400
CAS Number: 838-96-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR