4-Acetamido-2-methylbenzoic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

A2838-25G A2838-5G
TCA2838-25GEA 415.54 CAD
TCA2838-25G TCA2838-5G
4-Acetamido-2-methylbenzoic acid ≥96.0% (by HPLC, titration analysis)
4-Acetamido-2-methylbenzoic acid
Formula: C₁₀H₁₁NO₃
MDL Number: MFCD02258874
CAS Number: 103204-69-9

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR