Tetradecylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: n-Tetradecylphosphonic acidTDPAN-tetradecylphosphonicacid1-Tetradecylphosphonic acid

T3183-5G T3183-1G
TCT3183-1GEA 127.98 CAD
TCT3183-1G TCT3183-5G
Tetradecylphosphonic acid ≥98.0% (by titrimetric analysis)
Tetradecylphosphonic acid
Formula: C₁₄H₃₁O₃P
MW: 278.37 g/mol
MDL Number: MFCD00015834
CAS Number: 4671-75-4
EINECS: 225-118-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR