Hexylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: n-Hexylphosphonic acid, HPA, N-hexylphosphonicacid

H1551-1G H1551-5G
TCH1551-1GEA 109.02 CAD
TCH1551-1G TCH1551-5G
Hexylphosphonic acid ≥98.0% (by titrimetric analysis)
Hexylphosphonic acid
Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
MDL Number: MFCD00013961
CAS Number: 4721-24-8
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR