Hexadecylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

H1488-1G H1488-5G
TCH1488-1GEA 171.9 CAD
TCH1488-1G TCH1488-5G
Hexadecylphosphonic acid ≥98.0% (by titrimetric analysis)
Hexadecylphosphonic acid
Formula: C₁₆H₃₅O₃P
MDL Number: MFCD00015836
CAS Number: 4721-17-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 94.0 to 98.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR