3-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3056-1G C3056-5G
TCC3056-1GEA 71.1 CAD
TCC3056-1G TCC3056-5G
3-Chloro-4-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-4-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MDL Number: MFCD04039888
CAS Number: 175883-60-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR