5-Bromo-2-methoxyphenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
B5173-1G
B5173-5G
TCB5173-1GEA
45.82
CAD
TCB5173-1G
TCB5173-5G
5-Bromo-2-methoxyphenylboronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
5-Bromo-2-methoxyphenylboronic acid
Formula:
C₇H₈BBrO₃ Melting Pt: 131°C |
MDL Number:
MFCD00093409 CAS Number: 89694-45-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 95.0 to 108.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...