1,4-Dioxane-2,5-dione ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

G0467-25G G0467-5G
TCG0467-25GEA 200.58 CAD
TCG0467-25G TCG0467-5G
1,4-Dioxane-2,5-dione ≥98.0% (by GC, titration analysis)
1,4-Dioxane-2,5-dione
Formula: C₄H₄O₄
Boiling Pt: 130°C
Storage Temperature: Freezer
MDL Number: MFCD00081108
CAS Number: 502-97-6
EINECS: 207-954-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization back titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR