4-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1062-1G F1062-5G
TCF1062-1GEA 134.3 CAD
TCF1062-1G TCF1062-5G
4-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MDL Number: MFCD03788423
CAS Number: 179899-07-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR