3-Fluoro-5-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1063-1G F1063-5G
TCF1063-1GEA 170.64 CAD
TCF1063-1G TCF1063-5G
3-Fluoro-5-methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Fluoro-5-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MDL Number: MFCD07363780
CAS Number: 609807-25-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR