2-Phenyl-9,10-anthraquinone ≥98.0% (by GC)

Supplier: TCI America

P2588-1G P2588-5G
TCP2588-1GEA 106.08 CAD
TCP2588-1G TCP2588-5G
2-Phenyl-9,10-anthraquinone ≥98.0% (by GC)
2-Phenyl-9,10-anthraquinone
Formula: C₂₀H₁₂O₂
MDL Number: MFCD00559262
CAS Number: 6485-97-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR