Methylcyclopentadiene dimer (so called) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

M2705-100ML M2705-500ML
TCM2705-100MLEA 41.08 CAD
TCM2705-100ML TCM2705-500ML
Methylcyclopentadiene dimer (so called) stabilized
Methylcyclopentadiene dimer
Formula: C₁₂H₁₆
Flash Pt: 53°C
MDL Number: MFCD00082297
CAS Number: 26472-00-4
EINECS: 247-724-5
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR