7-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H1650-1G H1650-5G
TCH1650-1GEA 83.74 CAD
TCH1650-1G TCH1650-5G
7-Hydroxy-1-tetralone ≥98.0% (by GC, titration analysis)
7-Hydroxy-1-tetralone
Formula: C₁₀H₁₀O₂
Boiling Pt: 215°C
MDL Number: MFCD01312225
CAS Number: 22009-38-7

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR