7-Nitro-1-tetralone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,4-Dihydro-7-nitronaphthalen-1(2H)-one, 7-Nitro-3,4-dihydronaphthalen-1(2H)-one

N0945-1G N0945-5G
TCN0945-1GEA 64.78 CAD
TCN0945-1G TCN0945-5G
7-Nitro-1-tetralone ≥98.0% (by GC)
7-Nitro-1-tetralone
Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
MDL Number: MFCD00019661
CAS Number: 40353-34-2
EINECS: 254-887-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR