6-Amino-1-tetralone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Amino-3,4-dihydronaphthalen-1(2H)-one, 6-Amino-1,2,3,4-tetrahydronaphthalen-1-one

A2642-1G A2642-5G
TCA2642-1GEA 58.46 CAD
TCA2642-1G TCA2642-5G
6-Amino-1-tetralone ≥98.0% (by GC, titration analysis)
6-Amino-1-tetralone
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00099462
CAS Number: 3470-53-9

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR