7-Bromo-1-tetralone ≥95.0% (by GC)
Supplier: TCI America
B4545-1G
B4545-5G
TCB4545-1GEA
82.16
CAD
TCB4545-1G
TCB4545-5G
7-Bromo-1-tetralone ≥95.0% (by GC)
7-Bromo-1-tetralone
Formula:
C₁₀H₉BrO Boiling Pt: 157°C |
MDL Number:
MFCD02179287 CAS Number: 32281-97-3 |
Specification Test Results
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...