6-Bromo-1-tetralone ≥95.0% (by GC)

Supplier: TCI America
Warning

B4841-1G B4841-5G
TCB4841-1GEA 282.82 CAD
TCB4841-1G TCB4841-5G
6-Bromo-1-tetralone ≥95.0% (by GC)
6-Bromo-1-tetralone
Formula: C₁₀H₉BrO
Boiling Pt: 116°C
MDL Number: MFCD04114378
CAS Number: 66361-67-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR